Tisk…

Sodium-4-hydroxybenzenesulphonate dihydrate 97%

Dodavatel: Thermo Scientific
Varování

A14808.0E A14808.22 A14808.36
A14808.0EEA 10750 CZK
A14808.0E A14808.22 A14808.36
Sodium-4-hydroxybenzenesulphonate dihydrate 97%
Sodium-4-hydroxybenzenesulphonate dihydrate

Purity: 97% CAS No.: 10580-19-5 Molecular Formula: C6H5NaO4S.2H2O Formula Weight: 232.20 (196.17anhy) Odour: Odourless Melting Point: 300° Beilstein Reference: 4040891 MDL No.: MFCD00044734

Vzorec: C₆H₉NaO₆S
MW: 232,19 g/mol
Bod tání: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00150724
CAS číslo: 10580-19-5
EINECS: 212-550-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR