4-(p-Tolyl)butyric acid 97%
Dodavatel: Thermo Scientific
|
Varování
|
A14680.03
A14680.06
A14680.14
A14680.03EA
992
CZK
A14680.03
A14680.06
A14680.14
4-(p-Tolyl)butyric acid 97%
4-(p-Tolyl)butyric acid
Purity: 97%
CAS No.: 4521-22-6
Molecular Formula: C11H14O2
Formula Weight: 178.23
Melting Point: 55-59°
Beilstein Reference: 639705
MDL No.: MFCD00021786
Vzorec:
C₁₁H₁₄O₂ MW: 178,23 g/mol Bod tání: 55…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021786 CAS číslo: 4521-22-6 EINECS: 224-848-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...