Cinnamyl chloride 95% stabilised
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
3-Chloro-1-phenyl-1-propene
Stabilization:
Stabilised with sodium carbonate (Na₂CO₃)
Cinnamyl chloride is used in the enantioselective total synthesis of helioporins C and E, bioactive marine diterpenes
Stab. with sodium carbonate
Purity: 95%
CAS No.: 2687-12-9
Molecular Formula: C6H5CH=CHCH2Cl
Formula Weight: 152.62
Storage Temperature: 2°C to 8°C
Flash Point: 79°(174°F)
Refractive Index: 1.5845
Melting Point: 7°
Boiling Point: 86-87°/2mm
Beilstein Reference: 741974
UN No.: UN2927
MDL No.: MFCD00000986
Vzorec:
C₉H₉Cl Bod varu: 86…87 °C (2 mmHg) Bod tání: 7 °C Hustota: 1,096 Bod vzplanutí: 79 °C (174 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000986 CAS číslo: 2687-12-9 EINECS: 220-246-4 UN: 2927 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...