Tisk…

2-(Pyridin-4-yl)acetic acid hydrochloride ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A14348.06 A14348.14
A14348.06EA 2930 CZK
A14348.06 A14348.14
2-(Pyridin-4-yl)acetic acid hydrochloride ≥98%
2-(Pyridin-4-yl)acetic acid hydrochloride

Purity: 98+% CAS No.: 6622-91-9 Molecular Formula: C7H7NO2.HCl Formula Weight: 173.60 Storage Temperature: 2°C to 8°C Odour: Odourless Melting Point: ca 136° dec. Beilstein Reference: 3696648 MDL No.: MFCD00012827

Vzorec: C₇H₈ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00012827
CAS číslo: 6622-91-9
EINECS: 229-576-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR