Tisk…

ɑ-Bromocinnamaldehyde 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Bromo-3-phenyl-2-propenal, 2-Bromo-3-phenylacrylaldehyde, Alpha-Bromocinnamaldehyde

A14230.14 A14230.22
A14230.14EA 1060 CZK
A14230.14 A14230.22
ɑ-Bromocinnamaldehyde 98%
alpha-Bromocinnamaldehyde

Purity: 98% CAS No.: 5443-49-2 Molecular Formula: C9H7BrO Formula Weight: 211.06 Melting Point: 68-72° MDL No.: MFCD00006965

Vzorec: C₉H₇BrO
MW: 211,06 g/mol
Bod tání: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00006965
CAS číslo: 5443-49-2
EINECS: 226-637-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR