4-Fluoro-ɑ-methylstyrene 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Fluoro-4-(1-methylethenyl)benzene, 2-(4-Fluorophenyl)-1-propene
A14147.09
A14147.18
A14147.30
A14147.09EA
3070
CZK
A14147.09
A14147.18
A14147.30
4-Fluoro-ɑ-methylstyrene 95%
4-Fluoro-alpha-methylstyrene
Purity: 95%
CAS No.: 350-40-3
Molecular Formula: C9H9F
Formula Weight: 136.17
Storage Temperature: 2°C to 8°C
Flash Point: 53°(127°F)
Refractive Index: 1.5110
Boiling Point: 183°
Beilstein Reference: 2039696
UN No.: UN1993
MDL No.: MFCD00042297
Vzorec:
C₉H₉F MW: 136,17 g/mol Bod varu: 183 °C Hustota: 1,009 Bod vzplanutí: 53 °C (127 °F) |
MDL Number:
MFCD00042297 CAS číslo: 350-40-3 EINECS: 206-501-2 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...