Tisk…

Benzo[b]thiophene-1,1-dioxide 98%

Dodavatel: Thermo Fisher Scientific

A14063.06 A14063.14
A14063.06EA 3920 CZK
A14063.06 A14063.14
Benzo[b]thiophene-1,1-dioxide 98%
Benzo[b]thiophene-1,1-dioxide

Purity: 98% CAS No.: 825-44-5 Molecular Formula: C8H6O2S Formula Weight: 166.20 Melting Point: 137-138° Beilstein Reference: 116442 MDL No.: MFCD00052178

Vzorec: C₈H₆O₂S
Bod tání: 137…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00052178
CAS číslo: 825-44-5
EINECS: 212-544-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR