1,2-Diamino-4-chloro-5-fluorobenzene 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A13953.03
A13953.06
A13953.03EA
1640
CZK
A13953.03
A13953.06
1,2-Diamino-4-chloro-5-fluorobenzene 98%
1,2-Diamino-4-chloro-5-fluorobenzene
Purity: 98%
CAS No.: 139512-70-2
Molecular Formula: C6H6ClFN2
Formula Weight: 160.58
Melting Point: 110-114°
UN No.: UN2811
MDL No.: MFCD00042485
Vzorec:
C₆H₆ClFN₂ Bod tání: 110…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042485 CAS číslo: 139512-70-2 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...