Methyl-2-amino-3-hydroxypropanoate hydrochloride ≥98%
Dodavatel: Thermo Scientific
Synonyms:
Methyl DL-serinate hydrochloride, H-DL-Ser-OMe·HCl, 3-hydroxy-1-methoxy-1-oxopropan-2-aminium chloride, methyl 2-amino-3-hydroxypropanoate, DL-Serine Methyl Ester Hydrochloride, H-DL-Ser-OMe.HCl
A13904.09
A13904.18
A13904.30
A13904.09EA
1380
CZK
A13904.09
A13904.18
A13904.30
Methyl-2-amino-3-hydroxypropanoate hydrochloride ≥98%
Methyl-2-amino-3-hydroxypropanoate hydrochloride
Purity: 98+%
CAS No.: 5619-04-5
Molecular Formula: C4H9NO3.HCl
Formula Weight: 155.58
Storage Temperature: 2°C to 8°C
Melting Point: ca 134° dec.
Beilstein Reference: 6067970
MDL No.: MFCD00012593
Vzorec:
C₄H₁₀ClNO₃ MW: 155,578 g/mol Bod tání: ∼ 134 °C (rozkl.) |
MDL Number:
MFCD00012593 CAS číslo: 5619-04-5 EINECS: 227-047-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...