(+)-Rutin trihydrate 95%
Dodavatel: Thermo Fisher Scientific
A13570.14
A13570.22
A13570.36
A13570.14EA
1230
CZK
A13570.14
A13570.22
A13570.36
(+)-Rutin trihydrate 95%
(+)-Rutin trihydrate
Purity: 97%
CAS No.: 250249-75-3
Molecular Formula: C27H30O16.3H2O
Formula Weight: 664.58 (610.52anhy)
Melting Point: 184° dec.
Beilstein Reference: 75455
Optical Rotation: +9° (c=0.5 in ethanol)
MDL No.: MFCD00149490
Vzorec:
C₂₇H₃₀O₁₆·3H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149490 CAS číslo: 250249-75-3 EINECS: 205-814-1 Merck Index: 14,3802 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...