1,4-Bis(2-methylstyryl)benzene 99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Bis-MSB, 1,4-Di(2-methylstyryl)benzene, 1-Methyl-2-(2-(4-[2-(2-methylphenyl)ethenyl]phenyl)ethenyl)benzene, p-Bis(o-Methylstyryl) Benzene
Purity: 99%
CAS No.: 13280-61-0
Molecular Formula: C24H22
Formula Weight: 310.44
Odour: Odourless
Melting Point: 179-184°
Beilstein Reference: 2375845
MDL No.: MFCD00008529
Vzorec:
C₂₄H₂₂ MW: 310,44 g/mol Bod tání: 179…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008529 CAS číslo: 13280-61-0 EINECS: 236-285-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...