Tisk…

Piperine 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: (2E,4E)-5-(Benzo[d][1,3]dioxol-5-yl)-1-(piperidin-1-yl)penta-2,4-dien-1-one, 1-[5-(1,3-Benzodioxol-5-yl)-2,4-pentadienoyl]piperidine, 1-Piperoylpiperidine, 1-Piperylpiperidine

A13510.06 A13510.14 A13510.22
A13510.06EA 1200 CZK
A13510.06 A13510.14 A13510.22
Piperine 98%
Piperine

Purity: 98% CAS No.: 94-62-2 Molecular Formula: C17H19NO3 Formula Weight: 285.34 Melting Point: 128-133° Beilstein Reference: 90741 MDL No.: MFCD00005839

Vzorec: C₁₇H₁₉NO₃
MW: 285,34 g/mol
Bod tání: 128…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00005839
CAS číslo: 94-62-2
EINECS: 202-348-0
Merck Index: 14,7472

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR