Tisk…

2-Nonenoic acid 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A13228.06 A13228.14 A13228.22
A13228.06EA 924 CZK
A13228.06 A13228.14 A13228.22
2-Nonenoic acid 98%
2-Nonenoic acid

Purity: 98% CAS No.: 3760-11-0 Molecular Formula: C9H16O2 Formula Weight: 156.23 Flash Point: 110°(230°F) Refractive Index: 1.4620 Odour: Recognizable Boiling Point: 130-132°/2mm Beilstein Reference: 1721634 UN No.: UN3265 MDL No.: MFCD00014020

Vzorec: C₉H₁₆O₂
Bod varu: 130…132 °C (2 mmHg)
Hustota: 0,93
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014020
CAS číslo: 3760-11-0
EINECS: 223-171-5
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR