Tisk…

4-Acetoxycinnamic acid predominantly trans ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-(4-Acetoxyphenyl)acrylic acid

A12979.06 A12979.22
A12979.06EA 1360 CZK
A12979.06 A12979.22
4-Acetoxycinnamic acid predominantly trans ≥98%
4-Acetoxycinnamic acid

Purity: 98+% CAS No.: 15486-19-8 Molecular Formula: C11H10O4 Formula Weight: 206.20 Odour: Odourless Melting Point: 205-208° Beilstein Reference: 2645987 MDL No.: MFCD00016847

Vzorec: C₁₁H₁₀O₄
MW: 206,2 g/mol
Bod tání: 205…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00016847
CAS číslo: 15486-19-8
EINECS: 239-512-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR