Tisk…

Procion blue H-5R

Dodavatel: Thermo Scientific

A12719.14 A12719.22
A12719.14EA 1240 CZK
A12719.14 A12719.22
Procion blue H-5R
Procion blue H-5R

CAS No.: 12236-84-9 MDL No.: MFCD00070851

Storage Temperature: Ambient
MDL Number: MFCD00070851
CAS číslo: 12236-84-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR