4-Chlorobenzenesulphinic acid sodium salt hydrate 97%
Dodavatel: Thermo Scientific
A12583.06
A12583.14
A12583.06EA
1440
CZK
A12583.06
A12583.14
4-Chlorobenzenesulphinic acid sodium salt hydrate 97%
4-Chlorobenzenesulphinic acid sodium salt hydrate
Purity: 97%
CAS No.: 14752-66-0
Molecular Formula: C6H4ClNaO2S.xH2O
Formula Weight: 198.60(anhy)
Melting Point:
Vzorec:
C₆H₄ClNaO₂S·xH₂O MW: 198,60 g/mol (anhy) Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00035602 CAS číslo: 14752-66-0 EINECS: 238-811-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...