Tisk…

Magnesium lactate trihydrate ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

39369.22 39369.14
39369.22EA 4470 CZK
39369.22 39369.14
Magnesium lactate trihydrate ≥99%
Magnesium lactate trihydrate

White to off-white powder/lumps


Solubility: Soluble in water, Slightly soluble in alcohol

Vzorec: Mg(C₃H₅O₃)₂·3H₂O
Storage Temperature: Ambient
MDL Number: MFCD00156534
CAS číslo: 18917-93-6
Merck Index: 14,5672

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR