Methyl-3-(2-hydroxyphenyl)propionate 97%
Dodavatel: Thermo Fisher Scientific
A12219.06
A12219.14
A12219.06EA
1730
CZK
A12219.06
A12219.14
Methyl-3-(2-hydroxyphenyl)propionate 97%
Methyl-3-(2-hydroxyphenyl)propionate
Purity: 97%
CAS No.: 20349-89-7
Molecular Formula: C10H12O3
Formula Weight: 180.21
Melting Point: 40-44°
MDL No.: MFCD00067757
Vzorec:
C₁₀H₁₂O₃ MW: 180.21 g/mol Bod tání: 40…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067757 CAS číslo: 20349-89-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...