Tisk…

6-Mercaptopurine monohydrate 98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 6-Purinethiol monohydrate, 6-Mercaptopurine hydrate, 7H-Purine-6-thiol hydrate, 6-Thiohypoxanthine monohydrate

A12197.03 A12197.06 A12197.14
A12197.03EA 889 CZK
A12197.03 A12197.06 A12197.14
6-Mercaptopurine monohydrate 98%
6-Mercaptopurine monohydrate

Purity: 98% CAS No.: 6112-76-1 Molecular Formula: C5H4N4S.H2O Formula Weight: 170.19 (152.17anhy) Melting Point: ca 315° dec. Beilstein Reference: 4012091 MDL No.: MFCD03854445

Vzorec: C₅H₄N₄S·H₂O
MW: 170.19 g/mol (152.17 anhy)
Bod tání: ∼ 315 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD03854445
CAS číslo: 6112-76-1
EINECS: 200-037-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR