4-Azaxanthone 97%
Dodavatel: Thermo Fisher Scientific
A12045.03
A12045.06
A12045.03EA
1340
CZK
A12045.03
A12045.06
4-Azaxanthone 97%
4-Azaxanthone
Purity: 97%
CAS No.: 6537-46-8
Molecular Formula: C12H7NO2
Formula Weight: 197.20
Melting Point: 180-182°
Beilstein Reference: 8657
MDL No.: MFCD00052253
Vzorec:
C₁₂H₇NO₂ Bod tání: 180…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052253 CAS číslo: 6537-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...