(Triphenylphosphoranylidene)acetaldehyde, (max. 3% H₂O) 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-(Triphenylphosphoranylidene)acetaldehyde, (Formylmethylene)triphenylphosphorane
A11709.09
A11709.14
A11709.22
A11709.09EA
3100
CZK
A11709.09
A11709.14
A11709.22
(Triphenylphosphoranylidene)acetaldehyde, (max. 3% H₂O) 97%
(Triphenylphosphoranylidene)acetaldehyde
Purity: 97%
CAS No.: 2136-75-6
Molecular Formula: (C6H5)3P=CHCHO
Formula Weight: 304.33
Storage Temperature: 2°C to 8°C
Melting Point: 183-188°
Beilstein Reference: 523797
MDL No.: MFCD00006994
Vzorec:
C₂₀H₁₇OP Bod tání: 183…188 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006994 CAS číslo: 2136-75-6 EINECS: 218-375-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...