N-Acetyl-L-tyrosine ethyl ester monohydrate 97%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Ac-Tyr-OEt·H2O
A11577.06
A11577.14
A11577.06EA
1860
CZK
A11577.06
A11577.14
N-Acetyl-L-tyrosine ethyl ester monohydrate 97%
N-Acetyl-L-tyrosine ethyl ester monohydrate
Purity: 97%
CAS No.: 36546-50-6
Molecular Formula: C13H17NO4.H2O
Formula Weight: 269.30 (251.28anhy)
Storage Temperature: -30°C to -10°C
Melting Point: 80-81°
MDL No.: MFCD00149093
Vzorec:
C₁₃H₁₉NO₅ Bod tání: 80…81 °C |
MDL Number:
MFCD00149093 CAS číslo: 36546-50-6 EINECS: 212-663-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...