2-(1,3-Benzothiazol-2-yl)acetonitrile 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-(Benzo[d]thiazol-2-yl)acetonitrile
A11343.06
A11343.14
A11343.06EA
2050
CZK
A11343.06
A11343.14
2-(1,3-Benzothiazol-2-yl)acetonitrile 98%
2-(1,3-Benzothiazol-2-yl)acetonitrile
Purity: 98%
CAS No.: 56278-50-3
Molecular Formula: C9H6N2S
Formula Weight: 174.23
Melting Point: 99-103°
Beilstein Reference: 128460
UN No.: UN3439
MDL No.: MFCD00051633
Vzorec:
C₉H₆N₂S MW: 174,23 g/mol Bod tání: 99…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051633 CAS číslo: 56278-50-3 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...