L-Alanine ethyl ester hydrochloride ≥98%
Dodavatel: Thermo Fisher Scientific
Synonyms:
H-Ala-OEt.HCl, Ethyl (S)-2-Aminopropionate Hydrochloride, L-Ethyl alanate hydrochloride, (S)-2-Aminopropionic Acid Ethyl Ester Hydrochloride, H-Ala-OEt·HCl
A11271.06
A11271.14
A11271.06EA
1370
CZK
A11271.06
A11271.14
L-Alanine ethyl ester hydrochloride ≥98%
L-Alanine ethyl ester hydrochloride
Purity: 98+%
CAS No.: 1115-59-9
Molecular Formula: C5H11NO2.HCl
Formula Weight: 153.61
Melting Point: 74-78°
Beilstein Reference: 3594395
Optical Rotation: +2.9° (c=2.5 in water)
MDL No.: MFCD00063662
Vzorec:
C₅H₁₁NO₂ Bod tání: 74…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063662 CAS číslo: 1115-59-9 EINECS: 214-225-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...