1-Methyl-2-pyrrolecarboxaldehyde 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
N-Methylpyrrole-2-carboxaldehyde, 1-Methylpyrrole-2-aldehyde, 1-Methylpyrrole-2-carboxaldehyde, 1-Methylpyrrol-2-carbaldehyde, 2-Formyl-1-methylpyrrole
Purity: 98%
CAS No.: 1192-58-1
Molecular Formula: C6H7NO
Formula Weight: 109.13
Storage Temperature: 2°C to 8°C
Flash Point: 72°(161°F)
Refractive Index: 1.5610
Boiling Point: 88-90°/22mm
Beilstein Reference: 107811
MDL No.: MFCD00003087
Vzorec:
C₆H₇NO MW: 109,13 g/mol Bod varu: 88…90 °C (22 mmHg) Hustota: 1,074 Bod vzplanutí: 72 °C (161 °F) |
MDL Number:
MFCD00003087 CAS číslo: 1192-58-1 EINECS: 214-755-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...