3,3'-Oxydipropionitrile 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Bis(2-cyanoethyl) Ether
A11005.09
A11005.18
A11005.09EA
1640
CZK
A11005.09
A11005.18
3,3'-Oxydipropionitrile 98%
3,3'-Oxydipropionitrile
Purity: 98%
CAS No.: 1656-48-0
Molecular Formula: O(CH2CH2CN)2
Formula Weight: 124.11
Flash Point:
Vzorec:
C₆H₈N₂O MW: 124.14 g/mol Bod varu: 119…121 °C (1 mmHg) Hustota: 1.043 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001960 CAS číslo: 1656-48-0 EINECS: 216-750-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...