Tisk…

3,4-Pyridinedicarboxylic acid anhydride 97%

Dodavatel: Thermo Scientific
Varování

A10771.14 A10771.03 A10771.06
A10771.14EA 14690 CZK
A10771.14 A10771.03 A10771.06
3,4-Pyridinedicarboxylic acid anhydride 97%
3,4-Pyridinedicarboxylic acid anhydride

Purity: 97% CAS No.: 4664-08-8 Molecular Formula: C7H3NO3 Formula Weight: 149.11 Melting Point: 75-79° MDL No.: MFCD00010680

Vzorec: C₇H₃NO₃
MW: 149,11 g/mol
Bod tání: 75…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00010680
CAS číslo: 4664-08-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR