2-Furoyltrifluoroacetone 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A10693.06
A10693.14
A10693.06EA
1560
CZK
A10693.06
A10693.14
2-Furoyltrifluoroacetone 98%
2-Furoyltrifluoroacetone
Purity: 98%
CAS No.: 326-90-9
Molecular Formula: C8H5F3O3
Formula Weight: 206.12
Flash Point: 87°(188°F)
Refractive Index: 1.5280
Melting Point: 20-21°
Boiling Point: 203°
Beilstein Reference: 168085
MDL No.: MFCD00020935
Vzorec:
C₈H₅F₃O₃ Bod varu: 203 °C Bod tání: 20…21 °C Hustota: 1,391 Bod vzplanutí: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00020935 CAS číslo: 326-90-9 EINECS: 206-315-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...