N-(tert-Butoxycarbonyl)-L-leucine hydrate 99%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Boc-L-Leucine, BOC-L-Leucine hydrate, Boc-Leu-OH hydrate, N-Boc-L-leucine hydrate
A10647.06
A10647.14
A10647.06EA
707
CZK
A10647.06
A10647.14
N-(tert-Butoxycarbonyl)-L-leucine hydrate 99%
N-(tert-Butoxycarbonyl)-L-leucine hydrate
Purity: 99%
CAS No.: 13139-15-6
Molecular Formula: C11H21NO4.xH2O
Formula Weight: 231.29(anhy)
Storage Temperature: 2°C to 8°C
Melting Point: 84-87°
Beilstein Reference: 1711814
Optical Rotation: -25° (c=2 in acetic acid)
MDL No.: MFCD00066067
Vzorec:
C₁₁H₂₁NO₄·xH₂O MW: 231.29 g/mol (anhy) Bod tání: 84…87 °C |
MDL Number:
MFCD00066067 CAS číslo: 13139-15-6 EINECS: 236-073-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...