Tisk…

Pivalamidine hydrochloride 98%

Dodavatel: Thermo Fisher Scientific
Varování

A10613.14 A10613.06
A10613.14EA 17230 CZK
A10613.14 A10613.06
Pivalamidine hydrochloride 98%
Pivalamidine hydrochloride

Purity: 98% CAS No.: 18202-73-8 Molecular Formula: C5H12N2.HCl Formula Weight: 136.63 Melting Point: 188-193° Beilstein Reference: 3618865 MDL No.: MFCD00051988

Vzorec: C₅H₁₂N₂
Bod tání: 188…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00051988
CAS číslo: 18202-73-8
EINECS: 242-091-1

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR