4-(4-Methylphenyl)-4-oxobutyric acid 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-Oxo-4-(p-tolyl)butanoic acid, 3-(4-Methylbenzoyl)propionic Acid, 4-(p-Tolyl)-4-oxobutyric Acid
A10290.06
A10290.14
A10290.22
A10290.06EA
1270
CZK
A10290.06
A10290.14
A10290.22
4-(4-Methylphenyl)-4-oxobutyric acid 98%
4-(4-Methylphenyl)-4-oxobutyric acid
Purity: 98%
CAS No.: 4619-20-9
Molecular Formula: C11H12O3
Formula Weight: 192.22
Melting Point: 124-128°
Beilstein Reference: 2099653
MDL No.: MFCD00020541
Vzorec:
C₁₁H₁₂O₃ MW: 192,21 g/mol Bod tání: 124…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00020541 CAS číslo: 4619-20-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...