Tisk…

2-Methyl-3-(2-furyl)propenal 97%

Dodavatel: Thermo Fisher Scientific
Varování

A10125.09 A10125.18
A10125.09EA 1290 CZK
A10125.09 A10125.18
2-Methyl-3-(2-furyl)propenal 97%
2-Methyl-3-(2-furyl)propenal

Purity: 97% CAS No.: 874-66-8 Molecular Formula: C8H8O2 Formula Weight: 136.15 Storage Temperature: 2°C to 8°C Flash Point: 100°(212°F) Refractive Index: 1.6140 Boiling Point: 280-281° Beilstein Reference: 2438 MDL No.: MFCD00063240

Vzorec: C₈H₈O₂
MW: 136.15 g/mol
Bod varu: 280…281 °C
Hustota: 1.097
Bod vzplanutí: >100 °C (212 °F)
MDL Number: MFCD00063240
CAS číslo: 874-66-8
EINECS: 212-866-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR