Tisk…

Nickel molybdenum oxide ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

89938.14 89938.22
89938.14EA 1890 CZK
89938.14 89938.22
Nickel molybdenum oxide ≥98%
Nickel molybdenum oxide

Purity: 98% CAS No.: 14177-55-0 Molecular Formula: NiMoO4 Formula Weight: 218.65 Physical Form: Powder Odour: Odourless MDL No.: MFCD00016255

Vzorec: NiMoO₄
MW: 218.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016255
CAS číslo: 14177-55-0
EINECS: 238-034-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR