Sodium isopropoxide
Dodavatel: Thermo Scientific
|
Nebezpečí
|
89446.06
89446.14
89446.22
89446.06EA
2230
CZK
89446.06
89446.14
89446.22
Sodium isopropoxide
Sodium isopropoxide
CAS No.: 683-60-3
Molecular Formula: C3H7NaO
Formula Weight: 82.08
Physical Form: Crystalline
Sensitivity: Air and Moisture Sensitive
Appearance: White
Melting Point: 70-75°
UN No.: UN3206
MDL No.: MFCD00061566
Vzorec:
C₃H₇NaO Bod tání: 70…75 °C Storage Temperature: Ambient |
MDL Number:
MFCD00061566 CAS číslo: 683-60-3 EINECS: 211-673-7 ADR: 4.2,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...