Tisk…

Aluminum di(isopropoxide)acetoacetic ester chelate (≥9,6% Al)

Dodavatel: Thermo Fisher Scientific
Varování

89350.22 89350.36
89350.22EA 2600 CZK
89350.22 89350.36
Aluminum di(isopropoxide)acetoacetic ester chelate (≥9,6% Al)
Aluminum di(isopropoxide)acetoacetic ester chelate

Purity: 9.6% min CAS No.: 14782-75-3 Molecular Formula: C12H23AlO5 Formula Weight: 274.3 Flash Point: 50°(122°F) Physical Form: Liquid Density: 1.05 Sensitivity: Moisture Sensitive Odour: Alcohol-like UN No.: UN1993 MDL No.: MFCD00135521

Vzorec: C₁₂H₂₃AlO₅
MW: 274.3 g/mol
Hustota: 1.05
Bod vzplanutí: 50 °C (122 °F)
Storage Temperature: Ambient
MDL Number: MFCD00135521
CAS číslo: 14782-75-3
EINECS: 238-851-7
ADR: 3 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR