Gadolinium(III) acetate hydrate ≥99.9%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
89158.06
89158.14
89158.22
89158.06EA
903
CZK
89158.06
89158.14
89158.22
Gadolinium(III) acetate hydrate ≥99.9%
Gadolinium(III) acetate hydrate
Purity: 99.9%
CAS No.: 100587-93-7
Molecular Formula: Gd(OOCCH3)3.xH2O
Formula Weight: 334.39(anhy)
Physical Form: Crystalline
Density: 1.611
Sensitivity: Hygroscopic
Odour: Odourless
Boiling Point: dec.
Solubility: Soluble in water
MDL No.: MFCD00150116
Vzorec:
Gd(OOCCH₃)₃·xH₂O Bod varu: Dec. Hustota: 1,611 Storage Temperature: Ambient |
MDL Number:
MFCD00150116 CAS číslo: 100587-93-7 EINECS: 240-204-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...