Amberlyst® 15 (H) ion exchange resin
Dodavatel: Thermo Scientific
Synonyms:
Strongly acidic macroreticular resin, Styrene-DVB
89079.18
89079.30
89079.A1
89079.18EA
1080
CZK
89079.18
89079.30
89079.A1
Amberlyst® 15 (H) ion exchange resin
Amberlyst® 15 (H)
CAS No.: 39389-20-3
Molecular Formula: Styrene-DVB; Strongly acidic macroreticular resin
Physical Form: Beads
Appearance: Pale brown
Odour: Odourless
MDL No.: MFCD00145841
Vzorec:
(C₁₀H₁₀·C₈H₈O₃S)x Storage Temperature: Ambient |
MDL Number:
MFCD00145841 CAS číslo: 39389-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...