Iron(II) gluconate hydrate ≥97%
Dodavatel: Thermo Fisher Scientific
88726.36
88726.A3
88726.36EA
1940
CZK
88726.36
88726.A3
Iron(II) gluconate hydrate ≥97%
Iron(II) gluconate hydrate
Purity: 97%
CAS No.: 22830-45-1
Molecular Formula: Fe[HOCH2(CHOH)4CO2]2.xH2O (x≈2)
Formula Weight: 446.19(anhy)
Physical Form: Granular
Melting Point: ca 140°
MDL No.: MFCD00150872
Vzorec:
C₁₂H₂₄FeO₁₅ Bod tání: ∼ 140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150872 CAS číslo: 22830-45-1 EINECS: 206-076-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...