Bronze (90:10 wt%) ≥99.0% (metals basis), powder -100 mesh
Dodavatel: Thermo Fisher Scientific
|
Varování
|
88359.30
88359.A1
88359.30EA
1540
CZK
88359.30
88359.A1
Bronze (90:10 wt%) ≥99.0% (metals basis), powder -100 mesh
Bronze
Purity: 99.0%
Molecular Formula: Cu:Sn; 90:10 wt%
Physical Form: Powder
Appearance: Grayish orange
Odour: Odourless
MDL No.: MFCD00198187