Iron nitride
Dodavatel: Thermo Scientific
88198.14
88198.22
88198.14EA
7280
CZK
88198.14
88198.22
Iron nitride
Iron nitride
CAS No.: 37245-77-5
Molecular Formula: FexN (x=2-4)
Physical Form: -325 Mesh Powder
MDL No.: MFCD00134717
Vzorec:
Fe_x N (x=2-4) Storage Temperature: Ambient |
MDL Number:
MFCD00134717 CAS číslo: 37245-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...