Tisk…

Hematoxylin hydrate, (max. 8% H₂O) ≥80% (dry weight), powder

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: (+)-Hematoxylin hydrate, (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol hydrate, Natural Black 1 hydtate, Hydroxybrasilin hydrate, (6aS,11bR)-7,11b-Dihydro-6H-indeno[2,1-c]chromene-3,4,6a,9,10-pentol hydrate

47223.14 47223.22
47223.14EA 3370 CZK
47223.14 47223.22
Hematoxylin hydrate, (max. 8% H₂O) ≥80% (dry weight), powder
Hematoxylin hydrate

Purity: 80% CAS No.: 517-28-2 Molecular Formula: C16H14O6.xH2O Formula Weight: 302.29(anhy) Physical Form: Powder Sensitivity: Light Sensitive Appearance: Cream to brown Melting Point: ca 200° dec. Beilstein Reference: 91399 MDL No.: MFCD00078111

Vzorec: C₁₆H₁₄O₆·xH₂O
MW: 320.3
Bod tání: ∼ 200 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS číslo: 517-28-2
EINECS: 208-237-3
Merck Index: 14,4637

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR