Tisk…

Scandium(III) bromide, ultra dry ≥99.99% (metals basis)

Dodavatel: Thermo Scientific


Ampuled under argon

47199.03 47199.06
47199.03EA 6320 CZK
47199.03 47199.06
Scandium(III) bromide, ultra dry ≥99.99% (metals basis)
Scandium(III) bromide

Purity: 99.99% CAS No.: 13465-59-3 Molecular Formula: ScBr3 Formula Weight: 284.67 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic MDL No.: MFCD00016324

Vzorec: Br₃Sc
MW: 284,67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016324
CAS číslo: 13465-59-3
EINECS: 236-699-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR