Tisk…

Dibismuth triselenide ≥99.995% (metals basis)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

47198.03 47198.06
47198.03EA 1630 CZK
47198.03 47198.06
Dibismuth triselenide ≥99.995% (metals basis)
Dibismuth triselenide

Purity: 99.999% CAS No.: 12068-69-8 Molecular Formula: Bi2Se3 Formula Weight: 654.84 Physical Form: -325 Mesh Powder Density: 6.82 Melting Point: 710° UN No.: UN3283 MDL No.: MFCD00014200

Vzorec: Bi₂Se₃
Bod tání: 710 °C
Hustota: 6,82
Storage Temperature: Ambient
MDL Number: MFCD00014200
CAS číslo: 12068-69-8
EINECS: 235-104-7
UN: 3283
ADR: 6.1,III
Merck Index: 14,1278

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR