Cerium tribromide, ultra dry ≥99.99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Cerous bromide, Cerium(III) bromide hydrate, Cerium (III) bromide
Ampuled under argon
47194.03
47194.06
47194.14
47194.03EA
2900
CZK
47194.03
47194.06
47194.14
Cerium tribromide, ultra dry ≥99.99% (metals basis)
Cerium tribromide
Purity: 99.99%
CAS No.: 14457-87-5
Molecular Formula: CeBr3
Formula Weight: 379.83
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 5.18
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 733°
Boiling Point: 1560°
MDL No.: MFCD00016004
Vzorec:
CeBr₃ MW: 379.83 Bod varu: 1560 °C Bod tání: 733 °C Hustota: 5.18 Storage Temperature: Ambient |
MDL Number:
MFCD00016004 CAS číslo: 14457-87-5 EINECS: 238-447-0 Merck Index: 14,1995 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...