Lutetium tribromide, ultra dry ≥99.99% (metals basis)
Dodavatel: Thermo Scientific
|
Varování
|
Ampuled under argon
47190.03
47190.06
47190.03EA
2640
CZK
47190.03
47190.06
Lutetium tribromide, ultra dry ≥99.99% (metals basis)
Lutetium tribromide
Purity: 99.99%
CAS No.: 14456-53-2
Molecular Formula: LuBr3
Formula Weight: 414.68
Physical Form: Powder, Ampouled under argon
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
MDL No.: MFCD00049965
Vzorec:
Br₃Lu MW: 414,68 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00049965 CAS číslo: 14456-53-2 EINECS: 238-446-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...