Lanthanum(III) iodide, ultra dry ≥99.99% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Lanthanum triiodide
Ampuled under argon
47184.03
47184.06
47184.03EA
2490
CZK
47184.03
47184.06
Lanthanum(III) iodide, ultra dry ≥99.99% (metals basis)
Lanthanum(III) iodide
Purity: 99.99%
CAS No.: 13813-22-4
Molecular Formula: LaI3
Formula Weight: 519.62
Physical Form: Powder, Ampouled under argon
Sensitivity: Moisture and Light Sensitive
Appearance: Off-white
Odour: Odourless
Melting Point: 772°
MDL No.: MFCD00049470
Vzorec:
I₃La Bod tání: 772 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049470 CAS číslo: 13813-22-4 EINECS: 237-465-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...