Aluminium bromide, ultra dry ≥99.999% (metals basis)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Aluminum bromide
Ampuled under argon
47168.06
47168.09
47168.06EA
8690
CZK
47168.06
47168.09
Aluminium bromide, ultra dry ≥99.999% (metals basis)
Aluminium bromide
Purity: 99.999%
CAS No.: 7727-15-3
Molecular Formula: AlBr3
Formula Weight: 266.71
Physical Form: Powder, Ampouled under argon
Density: 3.21
Sensitivity: Moisture Sensitive
Appearance: White to orange
Odour: Pungent
Melting Point: 97.5°
Boiling Point: 265°
UN No.: UN1725
MDL No.: MFCD00003421
Vzorec:
AlBr₃ MW: 266.69 Bod varu: 265 °C Bod tání: 97.5 °C Hustota: 3.21 Storage Temperature: Ambient |
MDL Number:
MFCD00003421 CAS číslo: 7727-15-3 EINECS: 231-779-7 UN: 1725 ADR: 8 II Merck Index: 14,332 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...