Tisk…

Manganese(II) iodide, ultra dry ≥99.99% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Manganous iodide

Ampuled under argon

47166.03 47166.06 47166.09
47166.03EA 4260 CZK
47166.03 47166.06 47166.09
Manganese(II) iodide, ultra dry ≥99.99% (metals basis)
Manganese(II) iodide

Purity: 99.99% CAS No.: 7790-33-2 Molecular Formula: MnI2 Formula Weight: 308.75 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 5.0 Sensitivity: Light, Air and Moisture Sensitive Appearance: White to pink to red Odour: Odourless Melting Point: 638° MDL No.: MFCD00016225

Vzorec: I₂Mn
Bod tání: 638 °C
Hustota: 5,0
Storage Temperature: Ambient
MDL Number: MFCD00016225
CAS číslo: 7790-33-2
EINECS: 232-201-6
Merck Index: 14,5732

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR