Tisk…

Neodymium triiodide, ultra dry ≥99.9% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Neodymium (III) iodide

Ampuled under argon

47157.03 47157.06
47157.03EA 4570 CZK
47157.03 47157.06
Neodymium triiodide, ultra dry ≥99.9% (metals basis)
Neodymium triiodide

Purity: 99.9% CAS No.: 13813-24-6 Molecular Formula: NdI3 Formula Weight: 524.95 Physical Form: Powder, Ampouled under argon Sensitivity: Moisture and Light Sensitive Appearance: Black Odour: Odourless MDL No.: MFCD00016248

Vzorec: I₃Nd
MW: 524.95
Bod tání: 784
Hustota: 5.85
Storage Temperature: Ambient
MDL Number: MFCD00016248
CAS číslo: 13813-24-6
EINECS: 237-467-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR