Tisk…

Iridium, černý ≥99.95% (metals basis)

Dodavatel: Thermo Fisher Scientific
Varování

47150.ME 47150.04 47150.09
47150.MEEA 14010 CZK
47150.ME 47150.04 47150.09
Iridium, černý ≥99.95% (metals basis)
Iridium

Purity: 99.95% CAS No.: 7439-88-5 Physical Form: S.A. 20-30m2/g Appearance: Black UN No.: UN3089 MDL No.: MFCD00011062

Vzorec: Ir
Storage Temperature: Ambient
MDL Number: MFCD00011062
CAS číslo: 7439-88-5
EINECS: 231-095-9
UN: 3089
ADR: 4.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR